Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABC540

Sigma-Aldrich

Anti-MEPE Antibody, clone LFMb-33

clone LFMb-33, from mouse

Synonym(s):

Matrix extracellular phosphoglycoprotein, Osteoblast/osteocyte factor 45, OF45, MEPE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LFMb-33, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MEPE(56955)

General description

The protein with the gene name MEPE and the protein name of Matrix extracellular phosphoglycoprotein or Osteoblast/osteocyte factor 45 (OF45) is an important secreted protein expressed by osteoblasts which promotes mineralization in bone. New research interestingly indicates that cancer cells that over express MEPE/OF45 show an increased resistance to radiation therapy, thus OF45 is being examined as a potentially new target for sensitizing tumor cells to radiochemotherapy. MEPE/OF45 is highly expressed by osteoblasts and osteocytes and my multiple cancer cell types and regulated by growth factors such as FGF-2 which acts to lower expression of the protein in normal cells.

Immunogen

Recombinant protein corresponding to human MEPE.

Application

Anti-MEPE Antibody, clone LFMb-33 | MABC540 is an antibody against MEPE Antibody for use in Western Blotting, Immunohistochemistry.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected MEPE in 10 µg of SK-N-SH and U2OS cell lysate.
Immunohistochemistry Analysis: A representative lot detected MEPE in human salivary gland tissue (Dent, J., et al. (2004). Research. 83(9):664-670).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected MEPE in 10 µg of HeLa cell lysate.

Target description

~65 kDa observed. The calculated molecular weight of this protein is 58 kDa, but can be observed at ~45-62 kDa due to high glycosylation.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service