Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC534

Sigma-Aldrich

Anti-TPM3 Antibody, isoform TC22, clone TC22-4

clone TC22-4, from mouse

Synonym(s):

Tropomyosin alpha-3 chain, Gamma-tropomyosin, Tropomyosin-3, Tropomyosin-5, hTM5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

TC22-4, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TPM3(7170)

General description

Tropomyosins are dimmers of coiled-coil proteins that provide stability to actin filaments and regulate access of other actin-binding proteins. The human TPM3 gene (Gene ID 7170; also known as TM3, TM5, TRK, CFTD, NEM1, TM-5, TM30, CAPM1, TM30nm, TPMsk3, hscp30, HEL-189, HEL-S-82p, OK/SW-cl.5) encodes seven tropomyosin alpha-3 chain isoforms (UniProt P06753; also known as Tropomyosin-3) as a result of alternative splicing. Spliced variants 1, 2, 4, and 5 are also known as the skeletal muscle, TM30nm, TC22, and hTM5 isoform, respectively. The TC22 and the hTM5 variants are identical in sequence other than the 25 amino acids at their C-terminal end. TC22 is expressed in 100% of colorectal carcinomas, but is not expressed in normal colon epithelial cells. The anticarcinogenic potency of sulfasalazine (SASP) and its active metabolite 5-aminosalicylate (5-ASA, mesalamine) in protecting ulcerative colitis (UC) patients from developing colorectal cancer is directly linked to their efficacy in reducing cellular expression of TC22. Likewise, siRNA-mediated TC22 downregulation is shown to significantly alter several critical carcinogenic pathways in the human colon adenocarcinoma LS180 cells.

Specificity

Clone TC22-4 selectively detects human tropomyosin alpha-3 chain (TMP3) spliced variant TC22 (isoform 4), but not spliced Isoform 1 (Skeletal muscle), 2 (TM30nm), 3, 5 (hTM5), 6, or 7.

Immunogen

Recombinant human TC22 and boosts with TC22-specific C-terminal peptide.

Application

This Anti-TPM3 Antibody, isoform TC22, clone TC22-4 is validated for use in Western blotting, Immunohistochemistry, ELISA and FLow Cytometry for the detection of TPM3.
Western Blotting Analysis: A representative lot detected endogenous TC22 in human colorectal carcinoma T84 cell lysate, as well as recombinant human TC22, but not five other human TPM3 isoforms (Lin, J.L., et al. (2002). Gastroenterology. 123(1):152-162; Das, K.K., et al. (2009). Mol Pharmacol. 76(1):183-191).
ELISA Analysis: A representative lot specifically detected recombinant human TC22, but not five other human TPM3 isoforms (Das, K.K., et al. (2009). Mol Pharmacol. 76(1):183-191.).
Flow Cytometry Analysis: A representative lot detected time-dependent TC22 downregulation in LS180 cells upon 2 mM 5-Aminosalicylate (5-ASA) treatment (Das, K.K., et al. (2009). Mol Pharmacol. 76(1):183-191.)
Immunohistochemistry: Representative lots detected TC22 expression in human colon carcinoma and adenomatous polyp tissues, but not in normal colon tissue samples (Lin, J.L., et al. (2002). Gastroenterology. 123(1):152-162; Watari, J., et al. (2008). Clin Gastroenterol Hepatol. 6(4):409-17).

Quality

Evaluated by Western Blotting in recombinant human TC22 protein.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected TPM3 in 10 µg of recombinant human TC22 protein.

Target description

~32 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service