Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC525

Sigma-Aldrich

Anti-BAG-1 Antibody, clone 3.10G3E2

clone 3.10G3E2, from mouse

Synonym(s):

BAG family molecular chaperone regulator 1, BAG-1, Bcl-2-associated athanogene 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3.10G3E2, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BAG1(573)

General description

BAG family molecular chaperone regulator 1 (BAG-1), also known as Bcl-2-associated athanogene 1, blocks a step in a pathway leading to apoptosis or programmed cell death. BAG-1 enhances the anti-apoptotic effects of BCL2 representing a link between growth factor receptors and anti-apoptotic mechanisms. BAG-1 is ubiquitously expressed throughout most tissues, except the liver, colon, breast, and uterine myometrium.

Specificity

This antibody detects 3 observed isoforms: BAG-1(S) at ~38 kDa, BAG-1 (M) at ~45 kDa & BAG-1 (L) at ~52 kDa.

Immunogen

GST-tagged recombinant protein corresponding to Human BAG-1.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-BAG-1 Antibody, clone 3.10G3E2 is validated for use in Western Blotting and Immunohistochemistry and Immunoprecipitation and ELISA for the detection of BAG-1.
Western Blot Analysis: A representative lot of this antibody detected BAG-1 in MCF-7 cell lysate (Brimmell, M., et al., (1999). British Journal of Cancer 81(5):1042–1051).

Western Blot Analysis: A representative lot of this antibody detected BAG-1 in colonic epithelial cell lysate (Arhel, N. J., et al., (2003). Int. J. Cancer: 106, 364–371).

Western Blot Analysis: A representative lot of this antibody detected BAG-1(L, M, S) in BAG-1(L, M, S) overexpressed MCF-7 cell lysates.

Immunohistochemistry Analysis: A representative lot of this antibody detected BAG-1 in breast cancer tissue sections (Cutress, R. I., et al., (2003). Oncogene. 22:4973–4982).

Immunohistochemistry Analysis: A representative lot of this antibody detected BAG-1 in breast cancer tissue sections.

Immunoprecipitation Analysis: A representative lot of this antibody immunoprecipitated BAG-1 from endogenouse MCF-7 cell lysates.

Immunoprecipitation Analysis: A representative lot of this antibody immunoprecipitated BAG-1 from Carcinoma-derived HT29 and transformed adenoma PC/AA/C1/SB10 cell lysates (Arhel, N. J., et al., (2003). Int. J. Cancer: 106, 364–371).

Interaction Assay/ELISA: A representative lot of this antibody detected BAG-1 by a BAG-1:HSC70 interaction/ELISA assay (Sharp, A., et al., (2009). FEBS Letters 583:3405–3411).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected BAG-1 in 10 µg of HeLa cell lysate.

Target description

~38, 45, & 52 kDa observed.
This antibody detects 3 observed isoforms at ~38 kDa (S), ~45 kDa (M) & ~52 kDa (L).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service