Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC1614M

Sigma-Aldrich

Anti-AATF Antibody, clone 1B2D8

clone 1B2D8, from rat

Synonym(s):

Protein AATF, Apoptosis-antagonizing transcription factor, Rb-binding protein Che-1, Traube protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B2D8, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... AATF(26574)
mouse ... Aatf(56321)

General description

Protein AATF (UniProt: Q9JKX4; also known as Apoptosis-antagonizing transcription factor, Rb-binding protein Che-1, Traube protein) is encoded by the Aatf (also known as Che1, Trb) gene (Gene ID: 56321) in murine species. AATF is a nuclear protein that functions as a general inhibitor of the histone deacetylase HDAC1. It binds to the pocket region of RB1 to displace HDAC1 from RB1/E2F complexes, leading to activation of E2F target genes and cell cycle progression. Conversely, displacement of HDAC1 from SP1 bound to the CDKN1A promoter leads to increased expression of this CDK inhibitor and blocks cell cycle progression. AATF is expressed in adrenal gland, Purkinje cells, heart, kidney, liver, lung, muscle, ovary, and testis. It is expressed uniformly throughout the embryo until E10.5. However, from E11.5, the relative expression level increases in the liver, hind brain, spinal cord, dorsal root ganglia, and the posterior commissure. Down-regulation of AATF is involved in colon carcinoma cell proliferation and over-expression of Aatf gene interferes with MAP3K12 induced apoptosis.

Specificity

Clone 1B2D8 is a rat monoclonal antibody that detects AATF in murine and human cells. It targets an epitope within the first 39 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to the first 39 amino acids from murine Apoptosis-antagonizing transcription factor (AATF).

application

Anti-AATF, clone 1B2D8, Cat. No. MABC1614, is a rat monoclonal antibody that detects AATF and it has been tested for use in Western Blotting and Immunocytochemistry.
Immunocytochemistry Analysis: A representative lot detected AATF in Immunocytochemistry applications (Ishigaki, S., et. al. (2010). Cell Death Differ. 17(5):774-86).

Western Blotting Analysis: A representative lot detected AATF in Western Blotting applications (Ishigaki, S., et. al. (2010). Cell Death Differ. 17(5):774-86).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected AATF in 10 µg of HeLa cell lysate.

Target description

~79 kDa observed; 59.48 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service