Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABC1203

Sigma-Aldrich

Anti-PFKFB3 Antibody, clone 4C10.2

clone 4C10.2, from mouse

Synonym(s):

6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 3, 6PF-2-K/Fru-2,6-P2ase 3, 6PF-2-K/Fru-2,6-P2ase brain/placenta-type isozyme, iPFK-2, PFK/FBPase 3, PFKFB3, Renal carcinoma antigen NY-REN-56

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4C10.2, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PFKFB3(5209)

General description

6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 3 (EC 2.7.1.105; EC 3.1.3.46; UniProt Q16875; also known as 6PF-2-K/Fru-2,6-P2ase 3, 6PF-2-K/Fru-2,6-P2ase brain/placenta-type isozyme, iPFK-2, PFK/FBPase 3, Renal carcinoma antigen NY-REN-56) is encoded by the PFKFB3 gene (Gene ID 5209) in human. The homodimeric bifunctional 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatases (PFKFBs) phosphorylate fructose 6-phosphate (F6P) to fructose-2,6-bisphosphate (F2,6BP), which in turn activates 6-phosphofructo-1-kinase and glycolytic flux to lactate. PFKFB3 is distinguished from the other three known PFKFBs (PFKFB1, 2, 4) by the presence of multiple copies of the AUUUA instability motif in its 3′-untranslated region, a very high kinase:phosphatase activity ratio (740:1), increased protein expression in rapidly proliferating transformed cells, solid tumors and leukemias, and regulation by several proteins essential for tumor progression. Heterozygous deletion results in reduced glucose metabolism and growth of Ras-transformed tumors in Pfkfb3+/- mice. PFKFB3 nuclear localization is reported in multiple cell lines due to the presence of a highly conserved C-terminal nuclear localization motif. F2,6BP is reported to enhance cyclin-dependent kinase (Cdk) activity toward p27 Thr187 phosphorylation in vitro, while ectopic expression of PFKFB3 in the nucleus is found to stimulate cellular proliferation without affecting glycolysis, suggesting a novel role of PFKFB3-mediated F2,6BP in cell cycle regulation. The canonical form of human PFKFB3 is a 520-a.a. protein composed of an N-terminal 6-phosphofructo-2-kinase domain (a.a. 1-245) and a C-terminal fructose-2,6-bisphosphatase domain (a.a. 246-520).

Specificity

The immunogen corresponds the C-terminal fragment of human PFKFB3 spliced isoforms 1, 3, and 4 reported by UniProt (Q16875), cross-reactivity toward spliced isoform 2 is likely, but has not been confirmed.

Immunogen

Epitope: C-terminal region.
GST-tagged recombinant human PFKFB3 C-terminal fragment.

Application

Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected PFKFB3 in A431 cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Glucose/Glycogen Metabolism
This Anti-PFKFB3 Antibody, clone 4C10.2 is validated for use in Western Blotting, Immunocytochemistry for the detection of PFKFB3.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected PFKFB3 in 10 µg of HEK293 cell lysate.

Target description

~57 kDa observed. 59.61 kDa (isoform 1), 58,88 kDa (isoform 2), 57.29 kDa (isoform 3), 60.60 kDa (isoform 4) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service