Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC1167M

Sigma-Aldrich

Anti-P53DINP1 Antibody, clone F8

clone F8, from rat

Synonym(s):

TP53INP1, Tumor protein p53-inducible nuclear protein 1, Stress-induced protein, p53-dependent damage-inducible nuclear protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rat

antibody form

purified antibody

antibody product type

primary antibodies

clone

F8, monoclonal

species reactivity

human, mouse

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Tumor protein p53-inducible nuclear protein 1 (UniProt: Q96A56; also known as Stress-induced protein, p53-dependent damage-inducible nuclear protein 1, p53DINP1) is encoded by the TP53INP1 (also known as P53DINP1, SIP) gene (GeneID: 94241) in human. p53DINP1 is a ubiquitously expressed protein with anti-proliferative and pro-apoptotic properties and is involved in cell stress response. It shuttles between the nucleus and the cytoplasm, depending on cellular stress conditions. It acts as a dual regulator of transcription and autophagy. p53DINP1 acts as a positive regulator of autophagy and in response to cellular stress or upon activation of autophagy, it relocates to autophagosomes where it interacts with autophagosome-associated proteins GABARAP, GABARAPL1/L2, MAP1LC3A/B/C to regulate autophagy. p53DINP1 also acts as an antioxidant and plays a major role in p53/TP53-driven oxidative stress response. It has both a p53/TP53-independent intracellular reactive oxygen species (ROS) regulatory function and a p53/TP53-dependent transcription regulatory function. p53DINP1 is reported to positively regulate p53/TP53 and p73/TP73 and stimulate their capacity to induce apoptosis. In response to double-strand DNA breaks, it promotes p53/TP53 phosphorylation on Ser46 and subsequent apoptosis. It is shown to acts as a tumor suppressor by inducing cell death by an autophagy and caspase-dependent mechanism.

Specificity

Clone F8 detects P53DINP1 (TP53INP1) protein in various human cancer cells.

Immunogen

GST-tagged full length recombinant human Tumor protein p53-inducible nuclear protein 1.

Application

Anti-P53DINP1, clone F8, Cat. No. MABC1167, is a rat monoclonal antibody that detects Tumor protein p53-inducible nuclear protein 1 (p53DINP1) and is tested for use in ELISA and Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: A 1:62.5 dilution from a representative lot detected P53DINP1 in 10 µg of human brain tumor, breast cancer, lymphoma, and pancreatic cancer tissue lysates.

ELISA Analysis: A representative lot detected P53DINP1 in Elisa applications (Saadi, H., et. al. (2013). Results Immunol. 3:51-6).

Western Blotting Analysis: A representative lot detected P53DINP1 in Western Blotting applications (Al Saati, T., et. al. (2013). Am J Pathol. 182(6):1996-2004; Peuget, S., et. al. (2014). Cell Death Differ. 21(7):1107-18; Saadi, H., et. al. (2013). Results Immunol. 3:51-6; Seillier, M., et. al. (2012). Cell Death Differ. 19(9):1525-35).

Quality

Evaluated by Western Blotting in human colon tissue lysate.

Western Blotting Analysis: A 1:62.5 dilution of this antibody detected P53DINP1 in 10 µg of human colon tissue lysate.

Target description

~27 kDa observed; 27.37 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service