Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC1139

Sigma-Aldrich

Anti-HPCAL1 Antibody, clone 1E10

clone 1E10, from mouse

Synonym(s):

Hippocalcin-like protein 1, Calcium-binding protein BDR-1, HLP2, Visinin-like protein 3, VILIP-3

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$182.00
100 μG
$429.00

$182.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$182.00
100 μG
$429.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$182.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E10, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... HPCAL1(3241)

General description

Hippocalcin-like protein 1 (UniProt: P37235; also known as Calcium-binding protein BDR-1, HLP2, Visinin-like protein 3, VILIP-3) is encoded by the HPCAL1 (also known a BDR1) gene (Gene ID: 3241) in human. HPCAL1 is a lipid-anchor membrane protein of the visinin-like subfamily of neural calcium sensor and is involved in the calcium-dependent regulation of rhodopsin phosphorylation. It is structurally related to Ca2+-myristoyl switch proteins that contain four EF-hand motifs and a covalently attached N-terminal myristoyl group. HPCAL1 is also expressed in Purkinje cells of the cerebellum and hippocampus where it is believed to regulate synaptic plasticity relevant for learning and memory. Clone 1E10 specifically detects denatured HPCAL1 protein that is endogenously expressed in several pancreatic cell lines. (Ref.: Chen, Y., et al. (2014). Monoclonal Antibodies in Immunodiagnosis and Immunotherapy 33(1); 20-27; Li, C., et al. (2016). PLoS One 11(11); e0165921).

Specificity

Clone 1E10 specifically detects Hippocalcin-like protein 1 (HPCAL1) in human cells.

Immunogen

Purified native and denatured human HPCAL1.

Application

Detects Hippocalcin-like protein 1 using this mouse monoclonal Anti-HPCAL1, clone 1E10, Cat. No. MABC1139, tested for use in Western Blotting and Immunoprecipitation.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: A representative lot detected HPCAL1 in Western Blotting applications (Chen, Y., et. al. (2014). Monoclon Antib Immunodiagn Immunother. 33(1):20-7).

Immunoprecipitation Analysis: A representative lot detected HPCAL1 in Immunoprecipitation applications (Chen, Y., et. al. (2014). Monoclon Antib Immunodiagn Immunother. 33(1):20-7).

Quality

Evaluated by Western Blotting in Panc-1 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected HPCAL1 in Panc-1 cell lysate.

Target description

~22 kDa observed; 22.13 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service