Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABC1038

Sigma-Aldrich

Anti-NQO2 Antibody, clone 4A4.1

clone 4A4.1, from mouse

Synonym(s):

Ribosyldihydronicotinamide dehydrogenase [quinone], NRH dehydrogenase [quinone] 2, NRH:quinone oxidoreductase 2, Quinone reductase 2, QR2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$388.00

$388.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$388.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$388.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A4.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NQO2(4835)

General description

Ribosyldihydronicotinamide dehydrogenase [quinone] (EC 1.10.5.1; UniProt P16083; also known as NRH dehydrogenase [quinone] 2, NRH:quinone oxidoreductase 2, Quinone reductase 2, QR2) is encoded by the NQO2 (also known as NMOR2) gene (Gene ID 4835) in human. NQO1 (EC 1.6.5.2) and NQO2 (EC 1.10.5.1; formerly EC 1.10.99.2) constitute the quinone oxidoreductase family in human. NQO1 is involved in vitamin K metabolism and in reducing the cellular quinone concentration to prevent the build-up of cellular reactive oxygen species (ROS). Both enzymes contain a tightly bound FAD as the oxidant cofactor, but differ in their reductant cofactors utilization. While NQO1 can efficiently utilize NADH or NADPH, NQO2 exhibits only low activity in the presence of NADH or NADPH. Instead, non-physiological compounds, such as dihydronicotinamide riboside (NRH), N-methyldihydronicotinamide, and dihydrobenzylnicotamide, are reported to act as good reductant cofactors in NQD2-catalyzed reactions in vitro. NQO1 & NQO2 catalyzed the reduction of a wide variety of substrates, including quinones, nitro-compounds, and iron (III) ions. NQO1 is inhibited by the anticoagulant dicoumarol, while NQO2 is only weakly inhibited by dicoumarol and more strongly inhibited by resveratrol.

Specificity

Clone 4A4.1 recognizes an internal epitope within the N-terminal half of human Quinone reductase 2.

Immunogen

Epitope: Internal (N-terminal half).
GST-tagged recombinant human NQO2 N-terminal fragment.

Application

Anti-NQO2 Antibody, clone 4A4.1 is an antibody against NQO2 for use in Western Blotting, Immunohistochemistry.
Research Category
Apoptosis & Cancer
Research Sub Category
Oxidative Stress
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected NQO2 in 10 µg of Jurkat cell lysate.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected NQO2 in human kidney, liver, and cerebral cortex tissue sections.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected NQO2 in 10 µg of HeLa cell lysate.

Target description

~26 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service