Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB858/BD-1P2-K

Sigma-Aldrich

Mouse Anti-Respiratory Syncytial Virus (RSV), clone 133-1H

clone 133-1H, from mouse

Synonym(s):

Anti-RSV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

clone

133-1H, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunofluorescence: suitable

shipped in

wet ice

General description

This Anti-RSV Antibody, fusion protein, all type A, B strains, clone 133-1H is validated for use in ELISA, IF for the detection of Respiratory Syncytial Virus.

Physical form

Format: Purified

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service