Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB8430

Sigma-Aldrich

Anti-Enterovirus Antibody, 70/71 Blend, clone 74-5G and 422-8D-4C-4D

Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

422-8D-4C-4D, monoclonal
74-5G, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable

isotype

IgG

shipped in

wet ice

Specificity

Reacts with the Enterovirus 70 and 71.

Application

Anti-Enterovirus Antibody, 70/71 Blend, clone 74-5G & 422-8D-4C-4D is an antibody against Enterovirus for use in IF & IC.
IFA at 1:100.

Dilute with buffer pH 7.5-8.0 to desired working volume.

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
In a buffer consisting of 0.02M PB+0.25M NaCl, pH=7.6 with 0.1% Sodium Azide as a preservative.

Storage and Stability

Maintain at 2-8°C in convenient undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yangang Pan et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 4(9), 1600489-1600489 (2017-09-22)
Viral entry into the host cell is the first step of virus infection; however, its dynamic process via endocytosis remains largely elusive. Here, the force tracing technique and single particle simulation are combined to investigate the invagination of single human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service