Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB80144

Sigma-Aldrich

Anti-Newcastle Disease Virus Antibody

clone HN4a, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HN4a, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

General description

Newcastle Disease Virus (NDV) is a member of the paramyxovirus group of enveloped negative-stranded RNA viruses.

Specificity

Specific to HN glycoprotein

Immunogen

Intact UV-inactivated Newcastle Disease virus strain Autralia-Victoria.

Application

Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Newcastle Disease Virus Antibody is validated for use in IP, WB for the detection of Newcastle Disease Virus.

Physical form

Format: Purified
Liquid in 0.02M PB with 0.25M NaCl, pH 7.6. Contains 0.1% sodium azide.

Storage and Stability

Store at 2° to 8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaqing Liu et al.
Virology journal, 16(1), 164-164 (2019-12-29)
The paramyxovirus haemagglutinin-neuraminidase (HN) is a multifunctional protein that is responsible for attachment to receptors, removal of receptors from infected cells to prevent viral self-aggregation (neuraminidase, NA) and fusion promotion. It is commonly accepted that there are two receptor binding
Miaomiao Chi et al.
The Journal of general virology, 101(1), 5-20 (2019-11-09)
To gain insights into the role of the head-stalk linker region in the fusion triggering, we constructed mutants by deleting or substituting the linker region (115-NGAANNSG-122) of Newcastle disease virus (NDV) haemagglutinin-neuraminidase (HN) with the corresponding sequences of other paramyxoviruses.
Monoclonal antibodies to newcastle disease virus: delineation of four epitopes on the HN glycoprotein
Iorio, R M and Bratt, M A
Journal of virology, 48, 440-450 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service