Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB5586

Sigma-Aldrich

Anti-Cyclooxygenase-1 Antibody

Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity

mouse, human, rat, bovine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PTGS1(5742)

Specificity

Cyclooxygenase 1 (Cox-1).

Immunogen

Purified Sheep Cox-1.

Application

Anti-Cyclooxygenase-1 Antibody is an antibody against Cyclooxygenase-1 for use in WB, ELISA.
Research Category
Neuroscience
Research Sub Category
Neuroinflammation & Pain

Neuroinflammation & Pain
Western blot: 1:200-1:2,000 using ECL. The antibody reacts with the ~70 kDa protein in uninduced NIH/3T3 cells (2).

Immunohistochemistry: not tested. It is suggested that the antibody be tried at 5-10 μg/mL.

ELISA: 1,10,000-1:30,000.

Optimal working dilutions must be determined by end user.

Linkage

Replaces: AB5116

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 100 mM Tris, pH 7.5 with 0.05% sodium azide.

Storage and Stability

Maintain -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service