Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB505A

Sigma-Aldrich

Mouse Anti-Rat IgG2b γ2b chain Antibody, Alkaline Phosphatase

Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$446.00

$446.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

$446.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

monoclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

IgG is one of the most abundant protein in human serum. IgG is important for our defence against microorganisms and the molecules are produced by B-lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Specificity

The antibody reacts with the γ2b heavy chain (Fc) of rat IgG2b.

Immunogen

Epitope: IgG2b γ2b chain
Prepared from rat IgG2b γ2b chain.

Application

Detect Rat IgG2b γ2b chain using this Mouse anti-Rat IgG2b γ2b chain Antibody, Alkaline Phosphatase validated for use in ELISA.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Physical form

Affinity Chromatography
Purified by affinity chromatography. Liquid in 50mM Tris/1mM MgCl2/50% Glycerol, pH 8.0, containing 0.1% NaN3 as preservative.

Storage and Stability

Maintain refrigerated at 2°-8°C under sterile conditions for up to twelve months from date of receipt. For l;ong term storage, aliquot and store at -20°C. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service