Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAB4500-C

Sigma-Aldrich

Anti-B23 Antibody, clone 3C9, Ascites Free

clone 3C9, from mouse

Synonym(s):

Nucleophosmin, B23, NPM, Nucleolar phosphoprotein B23, Nucleolar protein NO38, Numatrin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C9, monoclonal

species reactivity

monkey, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NPM1(4869)

General description

Nucleophosmin (UniProt P06748; also known as B23, NPM, Nucleolar phosphoprotein B23, Nucleolar protein NO38, Numatrin) is encoded by the NPM1 (also known as NPM) gene (Gene ID 4869) in human. Nucleophosmin (NPM) is a nucleolar protein that plays a role in ribosome biogenesis, nucleocytoplasmic transport, centrosome duplication, embryonic development, histone chaperone function, and transcriptional regulation. NPM is phosphorylated by Cdk2-cyclin E on Thr199 which causes the dissociation of NPM from the centrosome to allow duplication during the cell cycle. NPM is phosphorylated by the mitotic Cdk1-cyclin B complex on Thr234/237 during mitosis, resulting in its reassociation with the duplicated centrosomes. The human NPM1 gene transcribes three alternatrively spliced isoforms, isoforms 1&2 contain a nucleolar localization signal at their C-terminal end (W288/290 of isoform 1 and W259/261 of isoform 2), which is missing in the spliced isoform 3. In addition, a nuclear export signal is introduced, rendering the aberrant cytoplasmic location of this spliced isoform (NPMc).

Specificity

Clone 3C9 detected the 37/43 kDa nucleolar protein B23 located in the nucleolar granules in various cells, most intensely in actively proliferating cell cultures (Bulycheva, T.I., et al. (2000). Klin. Lab. Diagn. (1):37-38).

Immunogen

Daudi nuclear extract.

Application

Anti-B23 Antibody, clone 3C9, Ascites Free is an antibody against B23 for use in Western Blotting, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.
Western Blotting Analysis: 1.0 µg/mL of this antibody from a representative lot detected B23 in 10 µg of HeLa and Jurkat cell lysates.
Immunocytochemistry Analysis: Representative lots immunostained the nucleoli of paraformaldehyde-fixed, Triton X-100-permeabilized human cells by fluorescent immunocytochemistry (JIn, Y.Q., et al. (2014). Cell Cycle. 13(10):1627-1638; Dergunova, N., et al. (2002). Immunol Lett. 83(1):67-72; Bulycheva, T.I., et al. (2000). Tsitologiia. 42(10):944-954; Bulycheva, T.I., et al. (2000). Klin. Lab. Diagn. (1):37-38).
Immunohistochemistry Analysis: A representative lot immunostained the nucleoli of dopaminergic (DA) neurons in formalin-fixed, paraffin-embedded human and mouse brain sections (Domanskyi, A., et al. (2011). FASEB J. 25(9):2898-2910; Rieker, C., et al. (2011). J. Neurosci. 12;31(2):453-460).
Immunoprecipitation Analysis: A representative lot immunoprecipitated endogenous B23/NPM and associated BRCA2 from COS-7 cell lysate (Wang, H.F., et al. (2011). Cancer Res. 71(1):68-77).
Immunoprecipitation Analysis: A representative lot immunoprecipitated B23 from HeLa cell lysates (Dergunova, N., et al. (2002). Immunol Lett. 83(1):67-72).
Western Blotting Analysis: A representative lot detected the 35-40 kDa B23 doublet bands in HeLa and Ramos cell lysates as well as in HeLa B23 immunoprecipitate obtained with clone 3C9 (Dergunova, N., et al. (2002). Immunol Lett. 83(1):67-72).

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected B23 in 10 µg of HEK293 cell lysate.

Target description

~37 kDa observed.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service