Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB430-I

Sigma-Aldrich

Anti-Epithelium Endothelial Cells Antibody, clone PHM-5

clone 18.29 (PHM-5), from mouse

Synonym(s):

Glomerular endothelial cells, Glomerular epithelial cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$182.00
100 μG
$498.00

$182.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$182.00
100 μG
$498.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$182.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

18.29 (PHM-5), monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

target post-translational modification

unmodified

Related Categories

General description

The glomeruli network of capillaries filters the blood and the filtrate then enters the renal tubule. Unlike most capillary beds, the glomerular capillaries exit into efferent arterioles rather than venules. The resistance that exists in efferent arterioles causes sufficient hydrostatic pressure within the glomerulus to provide the force for ultrafiltration. Glomerular endothelial cells that create an interface between blood and tissue compartments are responsible for regulating vasomotor tone, trafficking of leukocytes, and maintenance of maintain hemostasis. Unlike endothelium in other tissues, the normal glomerular endothelium is extremely attenuated, being only about 50 to 150 nm in height around the perimeter of the capillary loops. Glomeruli of human kidneys are shown to be rich sialoglycoprotein, which is chemically distinct from rat podocalyxin. (Ref.: Ballermann, BJ (2005). Kidney Int. 67(5); 1668-1671; Kerjaschki, D., et al. (1986). J. Clin. Invest. 78(5); 1142-1149).

Specificity

Clone PHM-5 is a mouse monoclonal antibody that detects human epithelium/endothelial cells.

Immunogen

Human Glomerular cells.

Application

Anti-Epithelium Endothelial Cells, clone PHM-5, Cat. No. MAB430-I, is a mouse monoclonal antibody that detects human human epithelium/endothelial cells and has been tested for use in Immunohistochemistry (Paraffin).
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Epithelium Endothelial Cells in human cerebral cortex tissue sections.
Research Category
Cell Structure

Quality

Evaluated by Immunohistochemistry (Paraffin) in human kidney tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected Epithelium Endothelial Cells in human kidney tissue sections.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service