Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB3773

Sigma-Aldrich

Anti-Annexin I Antibody, clone 6E4/3

clone 6E4/3, Chemicon®, from mouse

Synonym(s):

Lipocortin I, Calpactin II, Chromobindin 9, Phospholipase A2 Inhibitory Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6E4/3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ANXA1(301)

General description

Annexin I belongs to a family of Ca2+ dependent phospholipid binding proteins. It has been implicated in the regulation of phagocytosis, cell signaling and cell proliferation.

Specificity

Recognizes human Annexin I

Immunogen

Whole native annexin proteins contained in a mixture of membrane proteins from human bone tissue

Application

Anti-Annexin I Antibody, clone 6E4/3 detects level of Annexin I & has been published & validated for use in ELISA, IF & WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western blot

Immunofluorescence

ELISA

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M phosphate buffer, 0.25M NaCl, pH 7.6, with 0.1% sodium azide

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months from date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supawat Chatchen et al.
Nephron, 139(1), 70-82 (2018-02-07)
Previous studies showed that ceftriaxone can cause acute kidney injury (AKI) in the pediatric population. This study proposed a cellular model of crystalline nephropathy in ceftriaxone-associated AKI and explored the related pathophysiology by using a proteomic approach. Ceftriaxone was crystallized
Wanying Miao et al.
Journal of immunology (Baltimore, Md. : 1950), 204(6), 1486-1498 (2020-02-09)
Microglia play essential roles in neuroinflammatory responses after traumatic brain injury (TBI). Our previous studies showed that phenotypes of microglia, as well as infiltrating macrophages, altered at different stages after CNS injury, which was correlated to functional outcomes. IL-13 is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service