Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB3682

Sigma-Aldrich

Anti-Cyclin A Antibody, clone E67.1

ascites fluid, clone E67.1, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

E67.1, monoclonal

species reactivity

human, mouse, mink, bovine

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CCNA1(8900)
mouse ... Ccna1(12427)

Specificity

Cyclin A. MAB3682 is able to immunoprecipitate active cdk/cyclin A H1 kinase from a wide variety of species. Cyclin A protein is absent in cells prior to S-phase, during which its levels increase and peak. Cyclin A is involved in the regulation of the cell cycle and is essential for progression through S phase. Cyclin A is a marker for actively proliferating cells and for cells in S phase.

Immunogen

Bovine Cyclin A.

Application

Anti-Cyclin A Antibody, clone E67.1 is a high quality Mouse Monoclonal Antibody for the detection of Cyclin A & has been validated in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western blot

Immunoprecipitation

Optimal working dilutions must be determined by the end user.

Physical form

Ascites fluid. Liquid. Contains no preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Runx2 disruption promotes immortalization and confers resistance to oncogene-induced senescence in primary murine fibroblasts.
Kilbey, A; Blyth, K; Wotton, S; Terry, A; Jenkins, A; Bell, M; Hanlon, L; Cameron, ER; Neil, JC
Cancer Research null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service