Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB2143

Sigma-Aldrich

Anti-VCAM-1 Antibody, clone B-N8

clone B-N8, Chemicon®, from mouse

Synonym(s):

CD106

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

B-N8, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

B-N8 specifically recognizes human VCAM-1 [CD106]

Application

Immunohistochemistry: 0.5 μg/mL on acetone-fixed cryostat sections.

Flow cytometry: 1 μg per 10E6 cells (HUVEC)

Function studies: B-N8 can be used in studies of cell adhesion molecules.

ELISA

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-VCAM-1 Antibody, clone B-N8 is validated for use in FC, IH for the detection of VCAM-1.

Physical form

> 95% purified immunoglobulin as determined by SDS-gel electrophoresis prior to addition of stabilizer and preservative. Liquid in PBS.
Format: Purified

Storage and Stability

Maintain at +2-8°C upon date of receipt. For long-term storage, aliquot and store at -20°C. Avoid repeated Freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service