Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1969

Sigma-Aldrich

Anti-Integrin αVβ1 Antibody

CHEMICON®, mouse monoclonal, JBS5

Synonym(s):

VLA-5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Integrin α5β1 Antibody, clone JBS5, ascites fluid, clone JBS5, Chemicon®

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

JBS5, monoclonal

species reactivity

human, monkey

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ITGA5(3678)

General description

Integrins are a family of dimeric, transmembrane proteins that mediate cell-cell and extracellular matrix adhesion. Signals transduced by integrins play a role in many biological processes, including cell growth, differentiation, migration and apoptosis. The integrin family is composed of at least 15 alpha and 8 beta subunits that may form over twenty different alpha-beta non-covalently bound dimeric combinations on the cell surface. The alpha subunits all have some homology to each other, as do the beta subunits. Both of the subunits contribute to the binding of the ligand. Integrin alpha subunits contain seven weak sequence repeats in the N-terminal region, which may be important in ligand binding, and have been predicted to fold cooperatively into a single beta-propeller domain with seven beta-sheets. The alpha-3 subunit (CD49c) is highly concentrated in epithelial cells where it strongly adheres to Laminin-5 and Laminin-5 induced rapid adhesion can be blocked by antibodies against the alpha-3 integrin subunit. The alpha-3 subunit exists in two different splice variants, denoted as "A" and "B". The only difference that results from this differential splicing is a total change in the cytoplasmic domain, while the extracellular domain stays the same. Knock-out mice lacking this subunit show prenatal lethality and abnormalities in the kidneys.

Specificity

Reacts with the human integrin alpha5beta1.

Immunogen

Jurkat Cells

Application

Anti-Integrin α5β1 Antibody, clone JBS5 is an antibody against Integrin α5β1 for use in IH, IH(P) & IP.
Immunoprecipitation: 1-2 μg


Induction of homotypic aggregation: 100 pg/mL


Immunohistochemistry with paraformaldehyde fixation.


Paraffin embedded, PFA fixed tissue in monkey 1:50 with HIER {Qin et al., 2003}.


Inhibition of adhesion to fibronectin


Optimal dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Ascites fluid containing no preservatives.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Mouse 3T3 fibroblasts Skin (Basement Membrane)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuho Tanaka et al.
Arthritis research & therapy, 15(5), R127-R127 (2013-11-30)
Articular chondrocytes undergo an obvious phenotypic change when cultured in monolayers. During this change, or dedifferentiation, the expression of type I and type III procollagen is induced where normal chondrocytes express little type I and type III procollagen. In this
A biomimetic peptide fluorosurfactant polymer for endothelialization of ePTFE with limited platelet adhesion.
Larsen, CC; Kligman, F; Tang, C; Kottke-Marchant, K; Marchant, RE
Biomaterials null
Jean-Baptiste Vincourt et al.
Cancer research, 70(11), 4739-4748 (2010-05-13)
Chondrogenic tumors that exhibit benign or malignant behaviors synthesize variable amounts of cartilage-like extracellular matrix. To define the regulators of these phenotypes, we performed a proteomic comparison of multiple human chondrogenic tumors, which revealed differential accumulation of the C-propeptides of
Tasuku Ohno et al.
PloS one, 12(9), e0184825-e0184825 (2017-09-22)
Angiopoietin-like protein 2 (ANGPTL2) maintains tissue homeostasis by inducing inflammation and angiogenesis. It is produced in infiltrating immune cells or resident cells, such as adipocytes, vascular endothelial cells, and tumor cells. We hypothesized that ANGPTL2 might play an important role
Suzanne McFarlane et al.
Oncotarget, 6(34), 36762-36773 (2015-10-09)
CD44 expression is elevated in basal-like breast cancer (BLBC) tissue, and correlates with increased efficiency of distant metastasis in patients and experimental models. We sought to characterize mechanisms underpinning CD44-promoted adhesion of BLBC cells to vascular endothelial monolayers and extracellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service