Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1926

Sigma-Aldrich

Anti-Fibronectin Antibody, cell binding domain, clone P1H11

clone P1H11, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P1H11, monoclonal

species reactivity

rat, human, primate

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FN1(2335)
rat ... Fn1(25661)

Specificity

Human fibronectin (CCBD)

Immunogen

Epitope: cell binding domain
Human fibronectin

Application

Immunohistochemistry: for use on acetone or paraformaldehyde fixed tissue

Immunocytochemistry: for use on cells that secrete fibronectin

Immunoblotting

Immunoprecipitation

ELISA

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Fibronectin Antibody, cell binding domain, clone P1H11 is validated for use in ELISA, IP, WB, IC, IH for the detection of Fibronectin.

Physical form

Format: Purified
Purified immunoglobulin.

Storage and Stability

Store at +2 - 8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark-Oliver Diesner et al.
Biointerphases, 6(4), 171-179 (2012-01-14)
Collecting information at the interface between living cells and artificial substrates is exceedingly difficult. The extracellular matrix (ECM) mediates all cell-substrate interactions, and its ordered, fibrillar constituents are organized with nanometer precision. The proceedings at this interface are highly dynamic
A new mesenchymal stem cell (MSC) paradigm: polarization into a pro-inflammatory MSC1 or an Immunosuppressive MSC2 phenotype.
Waterman, RS; Tomchuck, SL; Henkle, SL; Betancourt, AM
Testing null
Gaëlle Villain et al.
The FEBS journal, 285(23), 4394-4412 (2018-10-20)
The extracellular matrix (ECM) is essential to provide mechanical support to tissues but is also a bioactive edifice which controls cell behavior. Cell signaling generated by ECM components through integrin-mediated contacts, modulates cell biological activity. In addition, by sequestrating or
Katja Vehanen et al.
Acta ophthalmologica, 86(1), 91-96 (2007-08-23)
The use of biopolymers in peribulbar injection for controlled drug delivery provides alternative options to eyedrops and intravitreal or surgical methods. Polymerizable biopolymers are especially likely to have a role because of their particular properties. In liquid form, they can
Karina H Nakayama et al.
Tissue engineering. Part A, 16(7), 2207-2216 (2010-02-17)
The goal of this study was the production of a decellularized kidney scaffold with structural, mechanical, and physiological properties necessary for engineering basic renal structures in vitro. Fetal, infant, juvenile, and adult rhesus monkey kidney sections were treated with either

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service