Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1923

Sigma-Aldrich

Anti-Tenascin Antibody, type III repeats 4 and 5, clone 4F8

clone 4F8, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

4F8, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TNC(3371)

Specificity

Binds to a recombinant fragment of human tenascin containing the fibronectin type III repeats, numbers 4 and 5.

Immunogen

Epitope: type III repeats 4 and 5
Purified human tenascin

Application

Detect Tenascin using this Anti-Tenascin Antibody, type III repeats 4 & 5, clone 4F8 validated for use in ELISA, WB & IC.
Immunocytochemistry at 2 g/mL on U251 cells

Immunoblotting (Western) at 2 g/mL to detect whole tenascin under reducing conditions

ELISA: 2 x 10-10 M endpoint dilution

Affinity constant Kd= 2 x 10-10

Final working dilutions must be determined by end user.

Physical form

Format: Purified
Liquid in 0.02M Phosphate, 0.25M Sodium Chloride pH 7.6,

with 0.1% Sodium Azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service