Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAB1682

Sigma-Aldrich

Anti-Actinin α Antibody, clone AT6/172

ascites fluid, clone AT6/172, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

AT6/172, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... ACTN1(87)

Specificity

Human alpha actinin. This antibody recognizes an epitope in the N-terminal region of alpha-actinin, probably between residues 1-24.

Immunogen

Epitope: alpha
Human platelet protein

Application

Anti-Actinin Antibody, α, clone AT6/172 detects level of Actinin & has been published & validated for use in IF, IP & WB.
Immunoblotting: ~100kDa on SDS reducing gels

Immunoprecipitation

Immunofluorescence: methanol fixed cells

Immunohistochemistry



Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Physical form

Liquid

Storage and Stability

Maintain frozen at -20°C for up to 12 months in undiluted aliquots. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa A McPherson et al.
DNA repair, 83, 102644-102644 (2019-07-18)
Cellular homeostasis is dependent on a balance between DNA damage and DNA repair mechanisms. Cells are constantly assaulted by both exogenous and endogenous stimuli leading to high levels of reactive oxygen species (ROS) that cause oxidation of the nucleotide dGTP
Myriam Jaraíz-Rodríguez et al.
Stem cell reports, 9(2), 451-463 (2017-07-18)
Connexin43 (CX43), a protein that forms gap junction channels and hemichannels in astrocytes, is downregulated in high-grade gliomas. Its relevance for glioma therapy has been thoroughly explored; however, its positive effects on proliferation are counterbalanced by its effects on migration
L Valmu et al.
European journal of immunology, 29(7), 2107-2118 (1999-07-31)
Leukocyte adhesion is a regulated process, which involves CD11/CD18 leukocyte integrins. CD11/CD18 acidity may be regulated intracellularly, and the CD18 polypeptide has previously been shown to become phosphorylated on serine and threonine after phorbol ester activation of T cells. Increased
Lin Ning et al.
Biology open, 4(2), 125-136 (2015-01-13)
ICAM-5 is a negative regulator of dendritic spine maturation and facilitates the formation of filopodia. Its absence results in improved memory functions, but the mechanisms have remained poorly understood. Activation of NMDA receptors induces ICAM-5 ectodomain cleavage through a matrix
Sharissa L Latham et al.
Nature communications, 9(1), 4250-4250 (2018-10-14)
Germline mutations in the ubiquitously expressed ACTB, which encodes β-cytoplasmic actin (CYA), are almost exclusively associated with Baraitser-Winter Cerebrofrontofacial syndrome (BWCFF). Here, we report six patients with previously undescribed heterozygous variants clustered in the 3'-coding region of ACTB. Patients present

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service