Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1669

Sigma-Aldrich

Anti-VEGF Receptor-2 Antibody, clone 89B3A5

clone 89B3A5, Chemicon®, from rat

Synonym(s):

FLK-1, KDR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

89B3A5, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

Specificity

FLK-1 is a receptor tyrosine kinase expressed on endothelial cells and on the common precursor of endothelial and hematopoietic stem cells Antibodies against FLK-1 are suitable for studies for endothelial development, for embryonal hematopoiesis, and for signal transduction via the FLK-1 receptor. It has not been tested whether the antibody blocks the binding of VEGF.

Immunogen

Rat L cells transfected with the complete coding sequence of mouse FLK-1 cDNA.

Application

Immunoblotting

Immunoprecipitation

FACS Analysis Optimal working dilutions must be determined by end user.
This Anti-VEGF Receptor-2 Antibody, clone 89B3A5 is validated for use in FC, IP, WB for the detection of VEGF Receptor-2.

Physical form

Format: Purified
Protein G purified immunoglobulin

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinhua Li et al.
Stem cells (Dayton, Ohio), 25(3), 697-706 (2006-12-16)
Recent evidence suggests that bone marrow (BM)-derived cells may integrate into the kidney, giving rise to functional renal cell types, including endothelial and epithelial cells and myofibroblasts. BM-derived cells can contribute to repair of the renal peritubular capillary (PTC) network

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service