Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAB16200-AF488

Sigma-Aldrich

Anti-MGMT Antibody, clone MT 3.1, Alexa Fluor 488 Conjugate

clone MT3.1, from mouse, ALEXA FLUOR 488

Synonym(s):

Methylated-DNA-protein-cysteine methyltransferase, 6-O-methylguanine-DNA methyltransferase, GATase, MGMT, O-6-methylguanine-DNA-alkyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

MT3.1, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MGMT(4255)

General description

Methylated-DNA-protein-cysteine methyltransferase (EC 2.1.1.63; UniProt P16455; also known as 6-O-methylguanine-DNA methyltransferase, GATase, MGMT, O-6-methylguanine-DNA-alkyltransferase) is encoded by the MGMT gene (Gene ID 4255) in human. MGMT is a DNA repair enzyme that removes alkyl groups from O6-guanine and plays a fundamental role in maintaining genomic integrity. MGMT converts the naturally occurring mutagenic DNA lesion O6-methylguanine back to guanine, thereby prevents mismatch and errors during DNA replication and transcription. MGMT expression level correlates with cellular resistance to alkylnitrosoureas, compounds that induce O6-alkylguanine lesions. MGMT transcription is negatively regulated via promoter methylation, MGMT promoter methylation level is reported to correclate with the survival rate of glioblastoma patients receiving treatment with alkylating agents.

Specificity

Clone MT 3.1 detected p53 expression-dependent MGMT downregulation in IMR90 cells (Harris, L.C., et al. (1996). Cancer Res. 56(9):2029-2032).

Application

Anti-MGMT Antibody, clone MT 3.1, Alexa Fluor 488 Conjugate, Cat. No. MAB16200-AF488, is a highly specific mouse monoclonal antibody that targets MGMT and has been tested in Flow Cytometry and Immunocytochemistry.
Flow Cytometry Analysis: 0.2 µL from a representative lot detected MGMT in one million MCF-7 cells.

This monoclonal antibody is also available in unconjugated and Alexa Fluor 647-conjugated forms (Cat. No. MAB16200 and MAB16200-AF647) for flow cytometry, immunocytochemistry, immunohistochemistry, immunoprecipitation, and Western blotting applications.

Quality

Evaluated by Immunocytochemistry in MCF-7 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this Alexa Fluor 488-conjugated antibody detected MGMT in MCF-7 cells.

Target description

21.65 kDa calculated. ~25 kDa reported (Brent, T.P., et al. (1990). Cancer Res. 50(1):58-61).

Physical form

Purified mouse IgG1 Alexa Fluor 488 conjugate in PBS with 1.5% BSA and 0.05% sodium azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service