Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1532

Sigma-Aldrich

Anti-Hexokinase Type I Antibody, a.a. 191-209, clone 21

ascites fluid, clone 21, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$498.00

$498.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$498.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$498.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

21, monoclonal

species reactivity

bovine, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Hk1(15275)
rat ... Hk1(25058)

Specificity

Recognizes a segmental epitope located between residues 191-209 of the Type I hexokinase sequence.

Immunogen

Epitope: a.a. 191-209
Rat Type I hexokinase.

Application

Anti-Hexokinase Type I Antibody, a.a. 191-209, clone 21 is an antibody against Hexokinase Type I for use in IC & WB.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Kinases & Phosphatases
Western blot: 1:500-1:2,000. Immunolocalization of Type I hexokinase: 1:500-1:2,000. Optimal working dilutions must be determined by end user.

Storage and Stability

Store at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Zhang et al.
Archives of medical science : AMS, 17(2), 406-416 (2021-03-23)
Progressive accumulation of amyloid-β (Aβ) is a pathological trait of Alzheimer's disease (AD). Amyloid-β increases free radical production in neuronal cells, leading to neuronal cell death. Hormone replacement therapy can reduce the incidence of AD, and oestrogen significantly improves the
José Carlos Valle-Casuso et al.
PloS one, 7(2), e32448-e32448 (2012-03-03)
In previous work we showed that endothelin-1 (ET-1) increases the rate of glucose uptake in astrocytes, an important aspect of brain function since glucose taken up by astrocytes is used to supply the neurons with metabolic substrates. In the present
Ana González-Sánchez et al.
Oncotarget, 7(31), 49819-49833 (2016-07-09)
Connexin43 (Cx43), the major protein forming gap junctions in astrocytes, is reduced in high-grade gliomas, where its ectopic expression exerts important effects, including the inhibition of the proto-oncogene tyrosine-protein kinase Src (c-Src). In this work we aimed to investigate the
Noriko Nakamura et al.
Biology of reproduction, 79(3), 537-545 (2008-05-30)
During epididymal transit, sperm acquire the ability to initiate rapid forward progressive motility on release into the female reproductive tract or physiological media. Glycolysis is the primary source of the ATP necessary for this motility in the mouse, and several

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service