Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

MAB1510

Sigma-Aldrich

Anti-Ubiquitin Antibody, clone Ubi-1 (aka 042691GS)

ascites fluid, clone Ubi-1 (042691GS), Chemicon®

Synonym(s):

Ubiquitin, ubiquitin-CEP80

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

Ubi-1 (042691GS), monoclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Ubiquitin is a highly conserved protein of about 8.5 kDa molecular weight, which has an ATP dependent role in the targeting of proteins for proteolytic degradation. To perform this function, the protein to be degraded is first covalently attached to the C terminus of ubiquitin, and the ubiquitinated complex is then recognized by a complex of degradative enzymes. Interestingly, ubiquitin also becomes covalently bonded to many types of pathological inclusions, which appear to be resistant to normal degradation. Therefore, ubiquitin antibodies are very useful for studies of these inclusions. For example, the neurofibrillary tangles and paired helical filaments diagnostic of Alzheimer′s disease, Lewy bodies seen in Parkinson′s disease, and Pick bodies found in Pick′s disease are all heavily ubiquitinated and can be readily visualized with ubiquitin antibodies.

Specificity

Expected to cross-react with Bovine, Chicken, and Drosophila.
Reactivity with other species has not been confirmed.
Ubiquitin (76 amino acid, 8.5 kDa) both conjugated and unconjugated. Shows staining of tangle material in Alzheimer affected brain, and also stains other pathological inclusions such as Lewy bodies (in Parkinson’s disease) and Pick bodies (in Pick’s disease).

Immunogen

Purified ubiquitin (bovine RBC) covalently coupled to KLH using glutaraldehyde.

Application

Immunohistochemistry (frozen or paraffin embedded sections):
1:250-1:1,000 dilution from a previous lot was used.

Electron Microscopy:
1:250-1:1,000 dilution from a previous lot was used.

Immunoprecipitation:
(Liang, 1995; Oberdorf, 1999)
Note: Suggested blocking agent is bovine serum albumin.

Immunoblotting:
1:250-1:1,000. The sensitivity of the assay can be enhanced by autoclaving the blot (Swerdlow, 1986).

Optimal working dilutions must be determined by end user.
Research Category
Protein Trafficking

Neuroscience
Research Sub Category
Ubiquitin & Ubiquitin Metabolism
This Anti-Ubiquitin Antibody, clone Ubi-1 (aka 042691GS) is validated for use in IH(P), IP, WB for the detection of Ubiquitin.

Quality

Routinely evaluated by Western Blot on Jurkat lysates.

Western Blot Analysis:
1:500 dilution of this lot detected UBIQUITIN on 10 μg of Jurkat3 lysates.

Target description

8.5 kDa

Linkage

Replaces: 04-454

Physical form

Mouse monoclonal IgG1κ in buffer containing containing 10 mM sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C in undiluted aliquotes from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Alzheimer′s diseased brain.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterogeneous inclusions in neurofilament inclusion disease.
Hirotake Uchikado, Ang Li, Wen-Lang Lin, Dennis W Dickson
Neuropathology : Official Journal of the Japanese Society of Neuropathology null
Analysis of polyubiquitin conjugates reveals that the Rpn10 substrate receptor contributes to the turnover of multiple proteasome targets.
Mayor, T; Lipford, JR; Graumann, J; Smith, GT; Deshaies, RJ
Molecular and Cellular Proteomics null
An inhibitor of a deubiquitinating enzyme regulates ubiquitin homeostasis.
Kimura, Y; Yashiroda, H; Kudo, T; Koitabashi, S; Murata, S; Kakizuka, A; Tanaka, K
Cell null
H Yamano et al.
The EMBO journal, 17(19), 5670-5678 (1998-10-02)
Programmed proteolysis of proteins such as mitotic cyclins and Cut2/Pds1p requires a 9-residue conserved motif known as the destruction box (D-box). Strong expression of protein fragments containing destruction boxes, such as the first 70 residues of Cdc13 (N70), inhibits the
Lithium treatment arrests the development of neurofibrillary tangles in mutant tau transgenic mice with advanced neurofibrillary pathology.
Leroy, Karelle, et al.
Journal of Alzheimer'S Disease, 19, 705-719 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service