Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAB1076-I

Sigma-Aldrich

Anti-DYNLT1 Antibody, clone 7A9F4H10

clone 7A9F4H10, from mouse

Synonym(s):

Dynein light chain Tctex-type 1, Protein CW-1, T-complex testis-specific protein 1 homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7A9F4H10, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DYNLT1(6993)

General description

Eukaryotic cells rely on actin and microtubule-based protein "motors" to generate intracellular movements. These motor proteins contain specialized domains that hydrolyze ATP to produce force and movement along a cytoskeletal polymer (microtubules in the case of the kinesin family and dyneins). In all cell types, dynein is a large protein complex composed of six distinct subunits and is responsible for most intracellular movement toward the minus ends of microtubules. Cytoplasmic dynein has many important roles in transporting membrane bound organelles, including endosomes, lysosomes, and mitochondria. It is also involved in the positioning of the centrosome, the nucleus, and the Golgi. Cytoplasmic dynein is also responsible for virus transport to the nucleus, retrograde axonal transport, and microtubule and neurofilament transport. Three families of dynein light chains (DYNLT) have been identified. Two isoforms of the DYNLT family of light chains, DYNLT1 and DYNLT3, have been proposed to link dynein to specific cargoes. Recent publications indicate that DYNLT1 binds to Rhodopsin and Doc2 whereas DYNLT3 does not.

Immunogen

Recombinant protein corresponding to human DYNLT1.

Application

Detect Dynein using this mouse monoclonal antibody, Anti-DYNLT1 Antibody, clone 7A9F4H10 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected DYNLT1 human kidney and lung tissues.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected DYNLT1 in 10 µg of HEK293 cell lysate.

Target description

~12 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxana Nekrasova et al.
Nature communications, 9(1), 1053-1053 (2018-03-15)
The epidermis is a multi-layered epithelium that serves as a barrier against water loss and environmental insults. Its morphogenesis occurs through a tightly regulated program of biochemical and architectural changes during which basal cells commit to differentiate and move towards

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service