MAB10226
Anti-Dengue Virus-2 Antibody, clone 3H5-1 | MAB10226
ascites fluid, clone 3H5-1, Chemicon®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
biological source
mouse
antibody form
ascites fluid
antibody product type
primary antibodies
clone
3H5-1, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
immunofluorescence: suitable
isotype
IgG1
shipped in
wet ice
General description
Dengue (DF) and dengue hemorrhagic fever (DHF) are caused by one of four closely related, but antigenically distinct, virus serotypes (DEN-1, DEN-2, DEN-3, and DEN-4), of the genus Flavivirus. each serotype is sufficiently different that there is no cross-protection and epidemics caused by multiple serotypes (hyperendemicity) can occur.
Specificity
The antibody is Dengue virus type 2 specific
Immunogen
Dengue virus type 2 antigens from infected mouse brains
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Andrea Troupin et al.
Journal of immunology (Baltimore, Md. : 1950), 197(11), 4382-4391 (2016-11-02)
Dengue virus (DENV) is a mosquito-borne flavivirus that causes serious global human disease and mortality. Skin immune cells are an important component of initial DENV infection and systemic spread. Here, we show that mast cells are a target of DENV
S-phase-dependent enhancement of dengue virus 2 replication in mosquito cells, but not in human cells.
Helt, Anna-Marija and Harris, Eva
Journal of virology, 79, 13218-13230 (2005)
Blocking the dengue virus 2 infections on BHK-21 cells with purified recombinant dengue virus 2 E protein expressed in Escherichia coli
Chiu, Mei-Wui and Yang, Yun-Liang
Biochemical and biophysical research communications, 309, 672-678 (2003)
Michael J Conway et al.
PLoS neglected tropical diseases, 10(9), e0004941-e0004941 (2016-09-16)
Aedes aegypti is the primary vector of several medically relevant arboviruses including dengue virus (DENV) types 1-4. Ae. aegypti transmits DENV by inoculating virus-infected saliva into host skin during probing and feeding. Ae. aegypti saliva contains over one hundred unique
Margaret E Ackerman et al.
Journal of immunological methods, 366(1-2), 8-19 (2011-01-05)
Phagocytosis can be induced via the engagement of Fcγ receptors by antibody-opsonized material. Furthermore, the efficiency of antibody-induced effector functions has been shown to be dramatically modulated by changes in antibody glycosylation. Because infection can modulate antibody glycans, which in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service