Pad traps limit cell movement in the Z-plane & barrier structures help retain cells within field of view.
Application
The M04T pad trap plates for 12 μm cells utilize microfabricated PDMS pillars that extend from the ceiling to trap suspension cells within a single focal plane for real time imaging & analysis
Legal Information
CELLASIC is a registered trademark of Merck KGaA, Darmstadt, Germany