Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LC0HCG6S2

Millipore

Millistak+® Lenticular Disc Filter

C0HC, filtration area 1.8 m²

Synonym(s):

Millistak+ Lenticular Disc Filters, C0HC media series, 16 in. diameter, 8-cell, silicone seal, 2/pack

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
$1,553.00

$1,553.00


Check Cart for Availability


Select a Size

Change View
2 EA
$1,553.00

About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690

$1,553.00


Check Cart for Availability

Product Name

Millistak+® Lenticular Disc Filter, C0HC, cellulose, filtration area 1.8 m2, hydrophilic

material

cellulose
filter aid (inorganic)
mixed cellulose esters (MCE)
polypropylene
silicone seal

Quality Level

description

Perfusion bioreactor fluid

feature

hydrophilic

manufacturer/tradename

Millistak+®

parameter

0.03 bar max. differential pressure (0.5 psid) (Reverse)
0.2 bar max. inlet pressure (3 psi) at 123 °C
0.21 bar max. differential pressure (3 psid) at 123 °C (Forward)
1 bar max. inlet pressure (15 psi) at 80 °C
1.03 bar max. differential pressure (15 psid) at 80 °C (Forward)
2.07 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. inlet pressure (30 psi) at 25 °C

diam.

40.64 cm (16 in.)

filtration area

1.8 m2

impurities

<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)

non-volatile residue (NVR)

1.50-2.02 mg/m2 (gravimetric analysis test)

total organic carbon (TOC) residue

416-593 ppb (gravimetric analysis test)
565-1037 ppb

input

sample type liquid

conductivity

5-7 μmho per cm at 25 °C (product water)

filter grade

C0HC

Looking for similar products? Visit Product Comparison Guide

General description

Device Configuration: Stack Disk
Stack Design: 16 in. diameter, 8-cell

Application

Perfusion bioreactor fluid

Specifications

Filter Grade: DE30 / DE60

Preparation Note

Sanitization Protocol
May be hot water sanitized for 20 cycles using purified water @ 80 °C for 30 minutes
Sterilization Method
May be steam sterilized for 3 steam-in-place (SIP) cycles of 30 minutes @ 123 °C

Analysis Note

Gravimetric Extractables: Post autoclave (1 cycle of 30 minutes at 123 °C) and pure water flush of 100 L/m2 (10 L/ft2) of media surface area
Product Water Conductivity at 25°C: after a pure water flush of 10 L/ft2 (100 L/m2) of media surface area per autoclaved (1 cycle of 30 minutes at 123 °C) 12" 16-stack device
The effluent of an autoclaved (1 cycle of 30 minutes @ 123 °C) stacked device is negative after a WFI flush of 10 liters per ft2 (100 L/m2) of media surface area
Total Organic Carbon (TOC) Residue: post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 10 liters per ft2 (100 L/m2) of media surface area

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service