Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L55DET6S1

Millipore

Millistak+® Lenticular Disc Filter

DE55, filtration area 3.47 m²

Synonym(s):

Millistak+ Lenticular Disc Filters, DE55 media series, 16 in. diameter, 16-cell, silicone seal

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,035.00

$1,035.00


Estimated to ship onJuly 08, 2025



Select a Size

Change View
1 EA
$1,035.00

About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690

$1,035.00


Estimated to ship onJuly 08, 2025


Product Name

Millistak+® Lenticular Disc Filter, DE55, cellulose, filtration area 3.47 m2, hydrophilic

material

cellulose
filter aid (inorganic)
polypropylene
silicone seal

Quality Level

description

For clarification of serum, plasma, vaccines, cell culture or other fluids.

feature

hydrophilic

manufacturer/tradename

Millistak+®

parameter

0.03 bar max. differential pressure (0.5 psid) (Reverse)
0.2 bar max. inlet pressure (3 psi) at 123 °C
0.21 bar max. differential pressure (3 psid) at 123 °C (Forward)
1 bar max. inlet pressure (15 psi) at 80 °C
1.03 bar max. differential pressure (15 psid) at 80 °C (Forward)
2.07 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. inlet pressure (30 psi) at 25 °C

H

27.6 cm (10.88 in.)

diam.

40.64 cm (16 in.)

filtration area

3.47 m2

impurities

<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)

non-volatile residue (NVR)

3.93-6.43 mg/m2 (gravimetric analysis test)

total organic carbon (TOC) residue

753-1,567 ppb (gravimetric analysis test)
753-1567 ppb

input

sample type liquid

conductivity

9-12 μmho per cm at 25 °C (product water)

filter grade

DE55

Looking for similar products? Visit Product Comparison Guide

General description

Device Configuration: Stack Disk
Stack Design: 16 in. diameter, 16-cell

Application

For clarification of serum, plasma, vaccines, cell culture or other fluids.

Specifications

Filter Grade: DE55

Preparation Note

Sanitization Protocol
May be hot water sanitized for 20 cycles using purified water @ 80 °C for 30 minutes
Sterilization Method
May be steam sterilized for 3 steam-in-place (SIP) cycles of 30 minutes @ 123 °C

Analysis Note

Gravimetric Extractables: Post autoclave (1 cycle of 30 minutes at 123 °C) and pure water flush of 50 L/m2 of media surface area
Product Water Conductivity at 25°C: after a pure water flush of 5 liters per ft2 (50 L/m2) of media surface area per autoclaved (1 cycle of 30 minutes at 123 °C) 12" 16-stack device
The effluent of an autoclaved (1 cycle of 30 minutes @ 123 °C) stacked device is negative after a WFI flush of 5 liters per ft2 (50 L/m2) of media surface area
Total Organic Carbon (TOC) Residue: ppb post autoclave (1 cycle of 30 minutes at 123 °C) and pure water flush of 5 liters per ft2 (50 L/m2) of media surface area

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service