Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L30CET6S1

Millipore

Millistak+® Lenticular Disc Filter

CE30, filtration area 3.47 m²

Synonym(s):

Millistak+ Lenticular Disc Filters, CE30 media series, 16 in. diameter, 16-cell, silicone seal

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$886.00

$886.00


Check Cart for Availability


Select a Size

Change View
1 EA
$886.00

About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690

$886.00


Check Cart for Availability

Product Name

Millistak+® Lenticular Disc Filter, CE30, cellulose, filtration area 3.47 m2, hydrophilic

material

cellulose
polypropylene
silicone seal

Quality Level

description

For clarification of serum, plasma, vaccines, cell culture or other fluids.

feature

hydrophilic

manufacturer/tradename

Millistak+®

parameter

1.03 bar max. differential pressure (15 psid) at 80 °C (Forward)
2.07 bar max. differential pressure (30 psid) at 25 °C (Forward)

H

27.6 cm (10.88 in.)

diam.

40.64 cm (16 in.)

filtration area

3.47 m2

impurities

<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)

total organic carbon (TOC) residue

1370-2120 ppb

input

sample type liquid

conductivity

8-10 μmho per cm at 25 °C (product water)

filter grade

CE30

Looking for similar products? Visit Product Comparison Guide

General description

Device Configuration: Stack Disk
Stack Design: 16 in. diameter, 16-cell

Application

For clarification of serum, plasma, vaccines, cell culture or other fluids.

Specifications

Filter Grade: CE30
Sodium: 3.45 - 4.76 mg/ft2 of Media

Calcium: 2.56 - 2.74 mg/ft2 of Media

Potassium: 0.17 - 0.91 mg/ft2 of Media

Magnesium: 1.19 - 2.03 mg/ft2 of Media

Iron: <0.1 mg/ft2 of Media

Lead: <0.1 mg/ft2 of Media

Aluminum: <0.1 mg/ft2 of Media

Titanium: <0.1 mg/ft2 of Media

Chromium: <0.1 mg/ft2 of Media

Manganese: <0.1 mg/ft2 of Media

Cobalt: <0.1 mg/ft2 of Media

Nickel: <0.1 mg/ft2 of Media

Copper: <0.1 mg/ft2 of Media

Zinc: <0.1 mg/ft2 of Media

Preparation Note

Sanitization Protocol
May be hot water sanitized for 20 cycles using purified water @ 80 °C for 30 minutes
Sterilization Method
May be steam sterilized for 3 steam-in-place (SIP) cycles of 30 minutes @ 123 °C

Analysis Note

Product Water Conductivity at 25°C: after a pure water flush of 5 liters per ft2 (50 L/m2) of media surface area per autoclaved (1 cycle of 30 minutes at 123 °C) 12" 16-stack device
The effluent of an autoclaved (1 cycle of 30 minutes @ 123 °C) stacked device is negative after a WFI flush of 10 liters per ft2 (100 L/m2) of media surface area
Total Organic Carbon (TOC) Residue: post autoclave (1 cycle of 30 minutes at 123 °C) and pure water flush of 5 liters per ft2 (50 L/m2) of media surface area

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service