for use with Immobilon Western HRP Substrates for use with Nitrocellulose for use with PVDF (Immobilon membranes) for use with TBS-T with NFDM for use with commercially available detection reagents
detection method
chemiluminescent colorimetric fluorometric
shipped in
ambient
storage temp.
room temp
General description
Immobilon® GO applies the principles of lateral flow assays for walk away processing of Western blots. Simply place your miniblot in the device and then add your wash and antibody solutions to the ports in the top. Return later to apply detection substrate and image your Western blot.
Application
For use on immunoblots, Western blots and dot blots.
Immobilon® GO applies the principles of lateral flow assays for walk away processing of Western blots. Simply place your miniblot in the device and then add your wash and antibody solutions to the ports in the top. Return later to apply detection substrate and image your Western blot.
Legal Information
Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A discussion of factors affecting observed vs. calculated molecular weight in gel electrophoresis and western blotting.
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.