Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IHCR2014-6

Sigma-Aldrich

IHC Select Anti-Neuron-Specific Enolase Antibody, prediluted, clone BBS/NC/VI-H14

clone BBS/NC/VI-H14, from mouse

Synonym(s):

NSE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

BBS/NC/VI-H14, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®
IHC Select

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ENO2(2026)

Specificity

Reacts with human gamma-gamma enolase (Mr 46000). This antibody does not cross react with alpha or beta subunits of NSE.

Immunogen

Gamma-gamma enolase purified from human brain

Application

Antibody is prediluted and ready to use for Immunohistochemistry of formalin-fixed, paraffin-embedded tissues.

Pretreatment: Heat Induced Epitope Retrieval (HIER). Recommend Citrate Buffer, pH 6.0 (Cat. No. 21545). No signal was detected without Epitope retrieval.

Incubation: 10 minutes with IHC Select Detection Kits.

NSE has been prediluted for use as the primary antibody with Chemicon′s IHC Select Detection Kits and Protocols (Catalog Nos. DAB050, DET-HP1000, APR050, and DET-APR1000), but other supplier′s IHC detection systems may be used. For optimized protocol details, visit www.chemicon.com and select the protocols link under Cat. No.IHCR2014-6.
This IHC Select Anti-Neuron-Specific Enolase Antibody, prediluted, clone BBS/NC/VI-H14 is validated for use in IH(P) for the detection of Neuron-Specific Enolase.

Physical form

Format: Purified
Monoclonal mouse antibody supplied in liquid format as tissue culture supernatant, diluted in PBS, pH 7.2 with stabilizers, 0.2% Tween 20, and 0.1% Kathon as a preservative.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service