Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HX0847

Supelco

Hydroxynaphthol Blue

Synonym(s):

Hydroxynaphthol Blue, calcium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H11N2Na3O11S3
CAS Number:
Molecular Weight:
620.47
UNSPSC Code:
12352107
EC Index Number:
264-197-7

form

powder

pH

2-3 (20 °C, 10 g/L in H2O)

solubility

340 g/L

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C20H14N2O11S3.3Na/c23-15-9-16(35(28,29)30)13-3-1-2-4-14(13)18(15)21-22-19-12-6-5-11(34(25,26)27)7-10(12)8-17(20(19)24)36(31,32)33;;;/h1-9,22,24H,(H,25,26,27)(H,28,29,30)(H,31,32,33);;;/q;3*+1/p-3/b21-18-;;;

InChI key

JQECMNGWLIFQLL-QRQZZYIYSA-K

Analysis Note

FormPowderLoss on drying (105°C)5% maxSuitability as metal indicatorTo pass test

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service