HTS216M
ChemiSCREEN Membrane Preparation Recombinant Human α1D Adrenergic Receptor with N-terminal truncation
Human alpha1D GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41106514
eCl@ss:
32161000
biological source
human
recombinant
expressed in Chem-1 cells
manufacturer/tradename
ChemiScreen
Chemicon®
technique(s)
ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... ADRA1A(148)
General description
The endogenous catecholamines epinephrine and norepinephrine have profound effects on smooth muscle activity, cardiac function, carbohydrate and fat metabolism, hormone secretion, neurotransmitter release, and central nervous system actions. These activities are mediated by GPCRs belonging to two subfamilies, the α- and β-adrenoceptors (Bylund et al., 1994). The three members of the α1 subclass of adrenoceptors, α1A, α1B and α1D, couple to Gq, and promote contraction of vascular and urinary tract smooth muscle, relaxation of intestinal smooth muscle, increased contractile force in the heart, and glycogenolysis and gluconeogenesis in the liver. The different subtypes have overlapping distributions and variably contribute to these effects depending on species and tissue. The α1D adrenergic receptor mediates smooth muscle contraction in several tissues. In the vasculature, activation of α1D increases blood pressure (Tanoue et al., 2002; Hosoda et al., 2005). In the urinary tract, α1D promotes bladder contraction. Antagonists of α1 receptors are used to treat bladder outlet obstruction, and this effect is thought to be mediated by α1D (Chen et al., 2005). The α1D adrenergic receptors has a relatively long N-terminal extracellular domain, and truncation of this domain has been shown to increase expression of the receptor at the cell surface (Pupo et al., 2003). Millipore′s α1D membrane preparations, which contain a version of α1D lacking residues 2-79, are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of agonists and antagonists of α1D. The membrane preparations exhibit a Kd of 0.4 nM for [3H]-prazosin. With 0.5 nM [3H]-prazosin, 5 µg/well α1D (Δ2-79) Membrane Prep typically yields greater than 5-fold signal-to-background ratio.
Truncated human ADRA1D cDNA encoding α1D lacking residues 2-79
Application
Radioligand binding assay and GTPγS binding.
Biochem/physiol Actions
GPCR Class: A
Protein Target: alpha1D
Target Sub-Family: Adrenergic
Quality
Table 1. Signal:background and specific binding values obtained in a competition binding assay with varying amounts of α1D (Δ2-79) receptor membrane prep.
SPECIFICATIONS: 1 unit = 5 µg membrane preparation
Bmax for [3H]-prazosin binding: 4.23 pmol/mg protein
Kd for [3H]-prazosin binding: ~0.4 nM
10 µg/well | 5 µg/well | |
---|---|---|
Signal:Background | 15.4 | 11.2 |
Specific Binding (cpm) | 778.8 | 670.5 |
SPECIFICATIONS: 1 unit = 5 µg membrane preparation
Bmax for [3H]-prazosin binding: 4.23 pmol/mg protein
Kd for [3H]-prazosin binding: ~0.4 nM
Specifications
Inucbation Conditions
RECOMMENDED ASSAY CONDITIONS: Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [3H]-prazosin. (PerkinElmer NET-823 )
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 5-fold signal:background with 3H labeled prazosin at 0.5 nM
RECOMMENDED ASSAY CONDITIONS: Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [3H]-prazosin. (PerkinElmer NET-823 )
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 5-fold signal:background with 3H labeled prazosin at 0.5 nM
Physical form
Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA no preservatives.
Packaging method: Membranes protein adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.
Packaging method: Membranes protein adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.
Storage and Stability
Maintain frozen at -70°C. Product is stable for at least 6 months from the date of receipt when stored as directed. Do not freeze and thaw.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Function of the lower urinary tract in mice lacking alpha1d-adrenoceptor.
Chen, Qin, et al.
The Journal of urology, 174, 370-374 (2005)
N-terminal truncation of human alpha1D-adrenoceptors increases expression of binding sites but not protein.
Pupo, Andre S, et al.
European Journal of Pharmacology, 462, 1-8 (2003)
K Horie et al.
British journal of pharmacology, 116(1), 1611-1618 (1995-09-01)
1. To investigate the structure-activity relationships of alpha-adrenoceptor agonists for the alpha 1-adrenoceptor subtypes, we have compared the imidazoline class of compounds, oxymetazoline and cirazoline, with the phenethylamine, noradrenaline, in their affinities and also in their intrinsic activities in Chinese
Akito Tanoue et al.
The Journal of clinical investigation, 109(6), 765-775 (2002-03-20)
To investigate the physiological role of the alpha(1D)-adrenergic receptor (alpha(1D)-AR) subtype, we created mice lacking the alpha(1D)-AR (alpha(1D)(-/-)) by gene targeting and characterized their cardiovascular function. In alpha(1D)-/- mice, the RT-PCR did not detect any transcript of the alpha(1D)-AR in
Chihiro Hosoda et al.
Molecular pharmacology, 67(3), 912-922 (2004-12-16)
To study the functional role of individual alpha1-adrenergic (AR) subtypes in blood pressure (BP) regulation, we used mice lacking the alpha1B-AR and/or alpha1D-AR with the same genetic background and further studied their hemodynamic and vasoconstrictive responses. Both the alpha1D-AR knockout
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service