Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HTS185RTA

Sigma-Aldrich

Ready-to-Assay EP2 Prostanoid Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$912.00

$912.00


Check Cart for Availability


Select a Size

Change View
1 KIT
$912.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.84

$912.00


Check Cart for Availability

Product Name

Ready-to-Assay EP2 Prostanoid Receptor Frozen Cells, Human EP2 GPCR frozen cells for Calcium Flux FLIPR Assays.

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

shipped in

dry ice

General description

Millipore’s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

Prostanoids bind to a family of 8 GPCRs to exert their biological effects (Narumiya and FitzGerald, 2001). The protanoid PGE2 causes pain, vasodilation, immunosuppression of T cells, bone resorption and promotion of carcinogenesis. Four related GPCRs, EP1, EP2, EP3 and EP4, each bind to PGE2, but the different G protein coupling status of each receptor leads to distinct biological effects. EP2 couples primarily to Gs to increase intracellular cAMP levels. Mice deficient in EP2 receptor showed impaired ovulation and fertilization, salt-sensitive hypertension (Kennedy et al., 1999). It has been shown that EP2 receptors are also involved in cancer associated immunodeficiency. Thus, genetic knockout of the EP2 receptor reduced tumor growth and prolonged survival in mice that had undergone isograft injection of MC26 or Lewis lung carcinoma cells (Yang et al., 2003). Millipore’s cloned human EP2-expressing cell line is made in the Chem-9 host, which supports high levels of recombinant EP2 expression on the cell surface and contains high levels of the promiscuous G protein to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists and antagonists at EP2.

Cell Line Description

GPCR Cell Lines
Host cells: Chem-9

Application

Human EP2 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

GPCR Class: A
Protein Target: EP2
Target Sub-Family: Prostanoid

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.

Fifty (50) mL of Media Component.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service