Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HTS090M

Sigma-Aldrich

ChemiScreen Recombinant Human OT Oxytocin Receptor Membrane Preparation

Human OT GPCR membrane prepation for Radioligand binding Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

biological source

human

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... OXTR(5021)

General description

Human Oxytocin Receptor
Oxytocin is a cyclic 9 amino acid peptide that differs from vasopressin in 2 amino acids. Despite the close similarities in sequence, oxytocin and vasopressin have different biological activities and bind to distinct G protein-coupled receptors. The oxytocin receptor, OT, couples primarily to Gq/11 to mobilize intracellular calcium. In female reproduction, oxytocin promotes uterine contraction and lactation; oxytocin is the most commonly used drug for induction of labor, whereas an oxytocin antagonist, atosiban, is under investigation to suppress preterm labor. Oxytocin/OT interaction in the CNS also plays an important role in stress, male and female sexual response, and sociality (Gimpl and Fahrenholz, 2001). Chemicon′s oxytocin membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of oxytocin receptor interactions with oxytocin. The membrane preparations exhibit a Kd of 0.53nM for [125I]-OVTA. With 5.0 ug/well Oxytocin Membrane Prep and 0.5 nM [125I]-OVTA, a greater than 6-fold signal-to-background ratio was obtained.

Biochem/physiol Actions

GPCR Class: A
Protein Target: OT
Target Sub-Family: Oxytocin

Quality

Signal:background and specific binding values obtained in a competition binding assay with varying amounts of Oxytocin Receptor membrane prep:
5 µg/well10 µg/well
Signal:Background 9 9
Specific Binding (cpm) 21871 24926


SPECIFICATIONS: 1 unit = 5 µg membrane preparation
Bmax: 4.5 pmol/mg
Kd: 0.53 nM

Specifications

Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I] OVTA (Perkin Elmer # NEX254)
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.

Physical form

One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 6-fold signal:background with 125I-labeled OVTA at 0.5 nM.

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membranes protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Gimpl et al.
Physiological reviews, 81(2), 629-683 (2001-03-29)
The neurohypophysial peptide oxytocin (OT) and OT-like hormones facilitate reproduction in all vertebrates at several levels. The major site of OT gene expression is the magnocellular neurons of the hypothalamic paraventricular and supraoptic nuclei. In response to a variety of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service