Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HTS000MC2

Sigma-Aldrich

ChemiSCREEN Chem-2 Membrane Preparation, Null Control

Chem-2 membrane preparation for Negative control in radioligand binding Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000

biological source

human

recombinant

expressed in Chem-2 cells

manufacturer/tradename

ChemiScreen
Chemicon®

concentration

0.5 mg/mL

technique(s)

radioligand binding assay (RLBA): suitable

shipped in

dry ice

General description

Chem-2 cells are a suspension cell line endogenously overexpressing Ga16 that is used as a host for Chemicon′s ChemiScreen GPCR Membrane Preparations. Chemicon′s ChemiScreen Wild-Type Chem-2 Membrane Preparations are ideal controls for validation of radioligand binding assays employing ChemiScreen GPCR Membrane Preparations in the Chem-2 host cell line.

Application

Negative control in radioligand binding assay.

Biochem/physiol Actions

GPCR Class: N/A
Protein Target: Null Control
Target Sub-Family: Control

Physical form

HOST CELLS: Chem-2TRANSFECTION: noneMembranes were adjusted to 0.5 mg protein/ml in 50 mM Tris pH 7.4, 10% glycerol and 1% BSA, rapidly frozen, and stored at -80°C. Use at the same protein concentration as the ChemiScreen GPCR Membrane Preparations of interest.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service