Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HKI3MAG-99K

Millipore

MILLIPLEX® Human Kidney Injury Magnetic Bead Panel 3 - Toxicity Multiplex Assay

The analytes available for this multiplex kit are: β-2-Microglobulin (β2M), Retinol Binding Protein 4 (RBP4), Uromodulin.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000

To order a Milliplex® kit, please search for your analyte of interest.

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 100-103%
sensitivity: 0.052-0.175 ng/mL
(MinDC = 2SD)

standard curve range: 0.05-50 ng/mL
(RBP4)

standard curve range: 0.10-100 ng/mL
(β-2-Microglobulin)

standard curve range: 0.10-100 ng/mL
(Uromodulin)

inter-assay cv: <15%
intra-assay cv: <10%

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Maintenance of physiological balance in the body is attributed in part to normal kidney function. Altered kidney function due to injury, drug toxicity or kidney failure may be life threatening. Toxicity is the leading cause of drug failure, so research is expanding in search of more sensitive, rapid methods to determine organ-specific damage as quickly as possible. Traditionally, blood urea nitrogen (BUN) and serum creatinine tests have been used to determine drug-induced damage to the kidney. These tests only detect kidney damage after it begins and do not allow for the elucidation of where in the kidney the damage has occurred. Specificity and earlier detection of kidney injury is vital, and this area of research is expanding in search of more sensitive, rapid methods for determining the specific area of damage in kidney.

The MILLIPLEX® portfolio provides valuable research assays to investigate multiple biomarkers of kidney injury in human urine samples using the Luminex® xMAP® instrument platform. The analytes available for this multiplex kit are: β-2-Microglobulin (β2M), Retinol Binding Protein 4 (RBP4), Uromodulin.

Panel Type: Toxicity

Application

  • Analytes: β-2-Microglobulin, RBP4, Uromodulin
  • Recommended Sample type:
  • Recommended Sample dilution: 1:500 in kit Assay Buffer. Customers need to determine the optimal dilution factor for their samples. Our recommendations are based on urine samples from normal subjects.
  • Assay Run Time: Overnight

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Other Notes

Sensitivity: Please see kit protocol for individual assay sensitivities.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

target_organs

Respiratory Tract

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Novel kidney toxicity biomarkers expand options for acute nephrotoxicity detection. Multiplex assays measure multiple renal damage biomarkers in small sample volumes, minimizing time and costs. See how MILLIPLEX® multiplex kidney toxicity assays detected vancomycin-induced subacute nephrotoxicity in rat models.

Multiplex toxicity assays enable researchers to simultaneously measure multiple toxicity biomarkers of various organs to gain a better understanding of toxicity. Explore how MILLIPLEX® multiplex toxicity assays are advancing research on liver injury, kidney toxicity, genotoxicity, and more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service