Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HAEP0470W

Millipore

6mm Hydrophobic Edge Membrane 0.45µm 47mm white plain

Filling machine operations, sterility testing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.22

material

mixed cellulose esters (MCE)
plain filter
white filter

sterility

non-sterile

feature

hydrophobic

packaging

box of

border W

6 mm , hydrophobic edge

filter diam.

47 mm

pore size

0.45 μm pore size

input

sample type bottled water
sample type sport drink
sample type wine
sample type cosmetics
sample type food(s)
sample type soft drink
sample type cider
sample(s) (environmental)
sample type beer
sample type beverage(s)
sample(s) (environmental)

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc
sterility testing

shipped in

ambient

General description

Filter Code: HAEP
Hydrophobic edge membranes are standard MF-Type filters with edges. The non-wetting edge prevents drug intrusion under holder sealing rim

Packaging

Blue box with 4 x 25 filters

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service