Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF240

Sigma-Aldrich

Sema3A Recombinant Protein

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$443.00

$443.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$443.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

$443.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

concentration

0.25 mg/mL

UniProt accession no.

shipped in

dry ice

General description

Semaphorins are a large family of well conserved proteins that participate in a variety of cell signaling functions. However, they are best known for their role in development and regeneration within the nervous system. These affects have been shown to be primarily mediated by the class 3 semaporins (Sema3). In particular, the Sema3A family member acts as both a chemorepellant for axonal growth and a chemoattractant for dendritic processes1,2.

Here we provide recombinant Sema3A protein to facilitate researchers studying neurite outgrowth. The protein produced is an Fc-chimera containing the highly conserved semphorin domain, the Ig-like domain, and the highly positively charged C-terminal region (without the 20 amino acids at the C-terminus)3.

Quality

Growth cone collapse testing was carried out to verify recombinant Sema3A protein induces collapse. Briefly, dorsal root ganglion cells were plated and allowed to grow overnight in differentiation media. The next day the media was replaced with differentiation media or differentiation media with 1µg/ml of Sema3A protein. The cells were then incubated for 30 minutes, fixed, and stained with FITC conjugated antibodies specific for phalloidin (Part No. 90228). Representative images of each assay condition are shown below:

Physical form

40µl of 0.25mg/ml recombinant protein.
Protein A affinity purified recombinant Sema3A protein in PBS buffer.

Storage and Stability

Store at -20°C or at -80°C for up to 6 months from date of receipt. Avoid repeated freeze-thaw cycles.

Analysis Note

The purity of the Sema3A protein was also determined by SDS-PAGE. Purified protein (2µg) was run using non-denaturing conditions and then coomassie blue stained. The protein detected was approximately 125kD in size.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lionel A T Meyer et al.
Methods in molecular biology (Clifton, N.J.), 1493, 171-183 (2016-10-28)
The axonal growth cone is a specialized structure enabling axon extension and proper guidance to its target by sensing the extracellular environment. A growth cone collapse assay is a popular approach designed to characterize the inhibitory effect of secreted guidance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service