Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FCMAB192F

Sigma-Aldrich

Milli-Mark® Anti-CD30-FITC Antibody, clone Ber-H2

clone Ber-H2, Milli-Mark®, from mouse

Synonym(s):

Tumor necrosis factor receptor superfamily member 8, CD30L receptor, Lymphocyte activation antigen CD30, Ki-1 antigen

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$301.00

$301.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 TESTS
$301.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

$301.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Ber-H2, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TNFRSF8(943)

General description

CD30 is a transmembrane cytokine receptor belonging to the tumor necrosis factor (TNF) receptor superfamily. Mature CD30 has a molecular mass of 120 kDa and is derived from a 90 kDa precursor protein (de Bruin, 1995). The extracellular domain of CD30 is homologous to that of TNF receptor superfamily members, whereas there is no homology in teh cytoplasmic domain, suggesting major differences in signaling mechanisms (Falini, 1995). CD30 expression is found on Hodgkin and Reed-Sternberg (H-RS) cells, and on activated B and T lymphocytes (de Bruin, 1995). The antibody from the Ber-H2 hybridoma cell line was included in the Fourth International Workshop on Human Leukocyte Differentiation Antigens and was found to belong to the CD30 cluster (Schwarting, 1989).

Specificity

Antibody recognizes Human CD30 antigen.

Immunogen

Co-cell line established from a human with Hodgkin′s disease of T-cell lineage.

application

Milli-Mark Anti-CD30-FITC Antibody, clone Ber-H2 detects level of CD30 & has been published & validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using Jurkat cells

Target description

120 kDa (de Bruin, 1995)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Jurkat cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CD 30 is a marker of undifferentiated human embryonic stem cells rather than a biomarker of transformed hESCs.
Maria A Lagarkova et al.
Cell cycle (Georgetown, Tex.), 7(22), 3610-3612 (2008-11-13)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service