Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

FCMAB108A4

Sigma-Aldrich

Anti-phospho-SMC1 (Ser957) Antibody, clone 5D11G5 Alexa Fluor 488

clone 5D11G5, from mouse, ALEXA FLUOR 488

Synonym(s):

CDLS2, DKFZp686L19178, DXS423E, KIAA0178, MGC138332, OTTHUMP00000061876, SB1.8, SMC1, SMC1A, SMC1L1, SMC1alpha, SMCB, Sb1.8, Smcb

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$419.00

$419.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 TESTS
$419.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$419.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5D11G5, monoclonal

species reactivity

human, Xenopus, bovine

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer957)

Gene Information

human ... SMC1A(8243)

General description

SMC (structural maintenance of chromosomes) proteins are Involved in chromosome cohesion during cell cycle and in DNA repair. ATM phosphorylates SMC1 on Ser957 and Ser966 both in vitro and in vivo. Phosphorylated SMC1 is part of the complex that functions as a downstream effector in both the ATM/NBS1 branch and the ATR/MSH2 branch of S-phase checkpoint.

Specificity

Antibody recognizes SMC1 phosphorylolated at Ser957.
Predicted to cross-react with xenopus and bovine based on sequence homology

Immunogen

Epitope: Phosphorylated at and around Ser957
KLH-conjugated, synthetic peptide corresponding to human SMC1 phosphorylated at Ser957

Application

Research Category
Epigenetics & Nuclear Function

Apoptosis & Cancer
Research Sub Category
Cell Cycle, DNA Replication & Repair
This Anti-phospho-SMC1 (Ser957) Antibody, clone 5D11G5 Alexa Fluor 488 is validated for use in WB, FC for the detection of phospho-SMC1 (Ser957).

Quality

Evaluated by Flow Cytometry with Hela Cells

Target description

150kDa Calculated

Physical form

Protein G Purified
Purified mouse monoclonal IgG1 conjugated to Alexa Flour 488 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 6 months from date of receipt. Protect from light.

Analysis Note

Control
HeLa Cells

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Flow cytometry dye selection tips match fluorophores to flow cytometer configurations, enhancing panel performance.

Troubleshooting guide offers solutions for common flow cytometry problems, ensuring improved analysis performance.

Protocols

Learn key steps in flow cytometry protocols to make your next flow cytometry experiment run with ease.

Explore our flow cytometry guide to uncover flow cytometry basics, traditional flow cytometer components, key flow cytometry protocol steps, and proper controls.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service