Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FCIM100158

FlowCellect Human CD4/CD8 T Cell Kit

This FlowCellect Human CD4/CD8 T Cell Assay provides a rapid & simple method to asses the percentage of CD4 & CD8 T cells in flow cytometry applications.

Synonym(s):

Flow Cellect

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NC.55

biological source

mouse

Quality Level

clone

monoclonal

species reactivity

human

manufacturer/tradename

FlowCellect

technique(s)

flow cytometry: suitable

detection method

fluorometric

shipped in

wet ice

General description

Millipore’s FlowCellect Human CD4/CD8 T Cell Assay provides a rapid and simple method to asses the percentage of CD4 and CD8 T cells. The CD8-FITC/CD4-PE/CD3-PECy5 cocktail consists of three anti-human antibodies CD3-PECy5, CD4-PE, and CD8-FITC which enables the researcher to obtain percentages of mature human T lymphocytes (CD3+),helper/inducer (CD3+CD4+) T lymphocytes, and suppressor/cytotoxic (CD3+CD8+)T lymphocytes in erythrocyte-lysed whole blood. The CD3 antibody Anti-CD3, UCHT1, reacts with the ε-chain of the CD3 part of the TCR/CD3 complex. CD3 is a pan-T marker expressed by normal and neoplastic T cells and uniquely allows the identification of all T cell lymphocytes. The CD4 antibody allows the identification of human helper/inducer CD4+ T cell (HLA Class II reactive) and recognizes a 60,000 Da surface antigen. Monocytes also express CD4 but at lower density, and have no co-expression of CD3 and hence can be distinguished away from CD4 T Cells in this kit. The CD8 antibody allows the identification of CD8, a 68 kDa transmembrane glycoprotein expressed by class I major histocompatibility complex restricted, mature suppressor/cytotoxic T cells, the great majority of cortical thymocytes and approximately 30% of medullary thymocytes. In addition a proportion of γδ T cells and NK cells express CD8. Inclusion of the CD3 antibody allows for the unique identification of the CD8 cytotoxic T Cells

The kit can thus distinguish both the CD4 T helper cells as well as the CD8 cytotoxic T Cells. The entire assay can be preformed in 35 minutes in a simple no wash manner thus allowing for percentages and absolute counts simultaneously when using the easyCyte HT System.
T cell lymphocytes (CD3+) constitute more than % of circulating lymphocytes and play a central role in both humoral and cell-mediated immunity. CD3 T cell can be further broadly classified as T helper lymphocytes that are CD3+CD4+ and cytotoxic T lymphocytes that are CD3+CD8+. Identification and enumeration of CD3 T lymphocytes, CD4 T lymphocytes and CD8 T lymphocytes is important in many immunological experiments that involve T cell characterization or study of T cell function. These studies may range from immunophenotyping experiments, studying T cells during development and disease or understanding impacts on T cells on treatment with specific compounds or mitogens or evaluating T cell specific responses. T lymphocytes (T cells) play critical roles in the regulation of immune responses, and are responsible for mediating many of the effector mechanisms of the immune system. Multiple studies have identified the identification and enumeration of CD4 T helper cells to be important in characterizing and monitoring immunodeficiency such as HIV, immunosuppression, and autoimmune diseases. Changes in T cell populations have also been implicated in chronic inflammation associated with the disease states such as cancer and atherosclerosis as well as in viral infection, bacterial infections, parasitic infections, sepsis, tuberculosis, burns, trauma, malnutrition, and stress. Similarly, identification and enumeration of CD3+, CD8+ populations are also useful in characterizing or monitoring autoimmune diseases and immune reactions such as transplant rejection, graft-versus-host disease and systemic lupus erythermatosus. People that have severe combined immunodefiencies or congenital or acquired immune defiencies often have an elevation in their CD8+ subset. A monitoring of both the CD4+ and CD8+ lymphocyte counts has also been be an important factor to predict the clinical prognosis of severe hepatitis patients.

Application

Research Sub Category
Immune Response

Toxicity

Autoimmune disease

Components

1. CD8-FITC/CD4-PE/CD3-PECY5 (Part No.4700-1355. One vial containing 1000 µL of Antibody Cocktail.

2. Guava 1X Lysing Solution (Part No. 4700-0082) One bottle containing 40 mL buffer.

Quality

Lot to lot in-process testing on all kit components

Legal Information

FlowCellect is a trademark of Luminex Corporation

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service