Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EX0539

Supelco

EDTA, Disodium Salt, Dihydrate

Synonym(s):

(Ethylenedinitrilo)tetraacetic Acid, (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt, Dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

pH

4-5 (20 °C, 50 g/L in H2O)

mp

110 °C (elimination of water of crystallisation)

solubility

100 g/L

storage temp.

room temp

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Appearance:white to almost white, crystalline powder
Assay (complexometric):99.0-101.0%
Heavy metals (as Pb):0.005% max
Identity (IR-spectrum):conforms
Insoluble matter:0.005% max
Iron (Fe):0.001% max
Nitrilotriacetic acid [(HOCOCH2)3 N]:0.1% max
pH (50 g/l; water):4.0-6.0

pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

pcodes

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service