Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DR1035

Sigma-Aldrich

Anti-SMG1 Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-ATM-Related Kinase, Anti-LIP, Anti-61E3.4, Anti-KIAA0421, Anti-ATX, Anti-PI-3-Kinase-Related Kinase SMG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMG1P5(595101)

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~340 kDa SMG protein.
Recognizes the ~340 kDa SMG protein in HeLa cell nuclear extracts.
This Anti-SMG1 Rabbit pAb is validated for use in Immunoblotting for the detection of SMG1.

Immunogen

Epitope: within amino acids 1625-1675
a synthetic peptide corresponding to amino acids 1625-167 of human SMG1

Application


Immunoblotting (0.1 g/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM NaCl, 10 mM phosphate, pH 7.2.

Other Notes

Abraham, R.T., 2004. DNA Repair (Amst.)3, 919.
Brumbaugh, K.M., et al. 2004. Mol. Cell.14, 585.
Yamashita, A., et al. 2001. Genes Dev.15, 2215.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madhuri Bhuvanagiri et al.
EMBO molecular medicine, 6(12), 1593-1609 (2014-10-17)
Nonsense-mediated RNA decay (NMD) is an RNA-based quality control mechanism that eliminates transcripts bearing premature translation termination codons (PTC). Approximately, one-third of all inherited disorders and some forms of cancer are caused by nonsense or frame shift mutations that introduce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service