Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CX0646

Sigma-Aldrich

Charcoal, Activated

4-12 Mesh

Synonym(s):

Charcoal, Activated, Carbon, Activated, Activated Carbon

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
UNSPSC Code:
11101522
EC Index Number:
231-153-3

form

granular

size

4-12 mesh

color

black

particle size

4-12 mesh (>=80%)

mp

3550 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Application

4-12 Mesh

Analysis Note

Adsorptive power: Report results (ml of 0.1N Na2S2O3)ml
Color: Black
Form: Granules
Mesh size (4-12): 80.0% min

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service