Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CX0175

Supelco

Calcium Chloride

Synonym(s):

desiccant

Sign Into View Organizational & Contract Pricing


About This Item

form

granular

Quality Level

pH

8-10 (20 °C, 100 g/L in H2O)

mp

772 °C

solubility

740 g/L

storage temp.

room temp

Application

anhydrous, 20 mesh

Analysis Note

Assay (complexometric):91.0% min
Color:White
Form:Granules
Mesh size (16-20):70% min

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service