Key Documents
CN1203001
Millipore
PP PLEAT CART 1.2UM 30C-0
Select a Size
1 EA
$391.00
$391.00
Check Cart for Availability
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
Change View
1 EA
$391.00
About This Item
UNSPSC Code:
23151806
eCl@ss:
32031690
$391.00
Check Cart for Availability
Notify Me
Get notified when this item is ready to ship via email.
material
polypropylene
Quality Level
pore size
1.2 μm nominal pore size
1.2 μm pore size
cartridge code
Code 0 (2-222; O-rings)
General description
Device Configuration: Cartridge